siRNA Details
| Gene Symbol: Fzd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008057.3 | NP_032083.3 | 1 | 2750 | ||
| GenBank mRNA | AK010239.2 | - | 1 | 331 | ||
| BC049781.1 | - | 1 | 2473 | |||
| BC063077.1 | - | 2 | 2081 | 6844727 | ||
More Information
Set Membership: |
|

Fzd7
Genomic Map