siRNA Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008066.3 | NP_032092.1 | 10 | 1756 | ||
| GenBank mRNA | AK039055.1 | - | 10 | 1757 | ||
| AK046953.1 | - | 5 | 1890 | |||
| AK048165.1 | - | 1654 | ||||
| BC115727.1 | - | 10 | 1678 | 40039253 | ||
| M86567.1 | - | 10 | 1507 | |||

Gabra2
Genomic Map