siRNA Details
| Gene Symbol: Gabrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038701.2 | NP_001033790.1 | 6, 7 | 741 | ||
| NM_008071.3 | NP_032097.1 | 6, 7 | 793 | |||
| GenBank mRNA | AK039070.1 | - | 6, 7 | 763 | ||
| AK083107.1 | - | 6, 7 | 742 | |||
| AK140718.1 | - | 2, 3 | 335 | |||
| AK147247.1 | - | 6, 7 | 747 | |||
| U14420.1 | - | 6, 7 | 774 | |||

Gabrb3
Genomic Map