siRNA Details
| Gene Symbol: Gabrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038701.2 | NP_001033790.1 | 8 | 1103 | ||
| NM_008071.3 | NP_032097.1 | 8 | 1155 | |||
| GenBank mRNA | AK039070.1 | - | 8 | 1125 | ||
| AK083051.1 | - | 8 | 1167 | |||
| AK083107.1 | - | 8 | 1104 | |||
| AK140718.1 | - | 4 | 698 | |||
| AK147247.1 | - | 8 | 1109 | |||
| U14420.1 | - | 8 | 1136 | |||

Gabrb3
Genomic Map