siRNA Details
| Gene Symbol: Gas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008086.2 | NP_032112.1 | 1 | 2181 | ||
| GenBank mRNA | BC055747.1 | - | 2 | 1276 | ||
| BC058628.1 | - | 2 | 1821 | 6401957 | ||
| X65128.1 | - | 1 | 2011 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gas1| Gene Symbol: Gas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008086.2 | NP_032112.1 | 1 | 2181 | ||
| GenBank mRNA | BC055747.1 | - | 2 | 1276 | ||
| BC058628.1 | - | 2 | 1821 | 6401957 | ||
| X65128.1 | - | 1 | 2011 | |||