siRNA Details
| Gene Symbol: Gas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008087.2 | NP_032113.1 | 2, 3 | 388 | ||
| GenBank mRNA | AK142903.1 | - | 4, 5 | 567 | ||
| BC013456.1 | - | 1, 2 | 156 | 4237356 | ||
| BC053446.1 | - | 2, 3 | 437 | 6336752 | ||
| M21828.1 | - | 2, 3 | 388 | |||

Gas2
Genomic Map