siRNA Details
| Gene Symbol: Gata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008093.2 | NP_032119.2 | 2 | 953 | ||
| GenBank mRNA | AK132020.1 | - | 2 | 974 | ||
| AK142213.1 | - | 2 | 955 | |||
| BC105653.1 | - | 3 | 666 | 40046623 | ||
| BC105654.1 | - | 3 | 727 | 40046622 | ||
| BC105655.1 | - | 3 | 576 | 40046621 | ||
| BC105656.1 | - | 3 | 232 | |||
| U84725.1 | - | 2 | 947 | |||

Gata5
Genomic Map