siRNA Details
| Gene Symbol: Gata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008093.2 | NP_032119.2 | 6 | 1760 | ||
| GenBank mRNA | AK132020.1 | - | 6 | 1781 | ||
| AK142213.1 | - | 6 | 1762 | |||
| U84725.1 | - | 6 | 1752 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gata5| Gene Symbol: Gata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008093.2 | NP_032119.2 | 6 | 1760 | ||
| GenBank mRNA | AK132020.1 | - | 6 | 1781 | ||
| AK142213.1 | - | 6 | 1762 | |||
| U84725.1 | - | 6 | 1752 | |||