siRNA Details
| Gene Symbol: CLEC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005127.2 | NP_005118.2 | 3 | 825 | ||
| GenBank mRNA | AB015628.1 | - | 3 | 318 | ||
| AK307334.1 | - | 3 | 352 | |||
| AK311232.1 | - | 2 | 245 | |||
| AK313916.1 | - | 3 | 329 | |||
| AY142147.1 | - | 3 | 302 | |||
| BC005254.1 | - | 3 | 319 | 3996700 | ||
| CB160326.1 | - | 1 | 48 | |||
| X96719.1 | - | 3 | 300 | |||

CLEC2B
Genomic Map