siRNA Details
| Gene Symbol: Gh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008117.3 | NP_032143.1 | 5 | 689 | ||
| GenBank mRNA | AK019952.1 | - | 5 | 604 | ||
| AK019954.1 | - | 5 | 604 | |||
| AK019956.1 | - | 5 | 601 | |||
| AK019959.1 | - | 5 | 604 | |||
| AK019961.1 | - | 5 | 604 | |||
| AK030419.1 | - | 5 | 656 | |||
| AK030715.1 | - | 5 | 604 | |||
| AK077488.1 | - | 3 | 280 | |||
| BC061157.1 | - | 5 | 603 | 30250975 | ||
| X02891.1 | - | 5 | 603 | |||

Gh
Genomic Map