siRNA Details
| Gene Symbol: Gif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008118.3 | NP_032144.2 | 9 | 1472 | ||
| GenBank mRNA | AK078933.1 | - | 9 | 1473 | ||
| BC118519.1 | - | 9 | 1458 | 40105765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gif| Gene Symbol: Gif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008118.3 | NP_032144.2 | 9 | 1472 | ||
| GenBank mRNA | AK078933.1 | - | 9 | 1473 | ||
| BC118519.1 | - | 9 | 1458 | 40105765 | ||