siRNA Details
| Gene Symbol: Gnaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008139.5 | NP_032165.3 | 7 | 1863 | ||
| GenBank mRNA | AK147393.1 | - | 7 | 1867 | ||
| BC057583.1 | - | 7 | 1604 | 6408959 | ||
| M55412.1 | - | 7 | 1405 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnaq| Gene Symbol: Gnaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008139.5 | NP_032165.3 | 7 | 1863 | ||
| GenBank mRNA | AK147393.1 | - | 7 | 1867 | ||
| BC057583.1 | - | 7 | 1604 | 6408959 | ||
| M55412.1 | - | 7 | 1405 | |||