siRNA Details
| Gene Symbol: Gp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008148.4 | NP_032174.2 | 2 | 445 | ||
| GenBank mRNA | AK031272.1 | - | 2 | 444 | ||
| AK172059.1 | - | 2 | 444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gp5| Gene Symbol: Gp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008148.4 | NP_032174.2 | 2 | 445 | ||
| GenBank mRNA | AK031272.1 | - | 2 | 444 | ||
| AK172059.1 | - | 2 | 444 | |||