siRNA Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008150.2 | NP_032176.2 | 1 | 228 | ||
| GenBank mRNA | BC006622.1 | - | 1 | 228 | 3967797 | |
| CJ187434.1 | - | 1 | 182 | |||
| X83577.1 | - | 1 | 549 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpc4| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008150.2 | NP_032176.2 | 1 | 228 | ||
| GenBank mRNA | BC006622.1 | - | 1 | 228 | 3967797 | |
| CJ187434.1 | - | 1 | 182 | |||
| X83577.1 | - | 1 | 549 | |||