siRNA Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008150.2 | NP_032176.2 | 3 | 786 | ||
| GenBank mRNA | BC006622.1 | - | 3 | 786 | 3967797 | |
| BQ559580.1 | - | 1 | 308 | |||
| CN710713.1 | - | 1 | 264 | |||
| X83577.1 | - | 3 | 1107 | |||

Gpc4
Genomic Map