siRNA Details
| Gene Symbol: Gpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008154.3 | NP_032180.1 | 2 | 1620 | ||
| GenBank mRNA | AK140302.1 | - | 2 | 1591 | ||
| AK140464.1 | - | 2 | 1574 | |||
| AK158917.1 | - | 2 | 1583 | |||
| D21062.1 | - | 1 | 1995 | |||
More Information
Set Membership: |
|

Gpr3
Genomic Map