siRNA Details
| Gene Symbol: Gpr33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008159.2 | NP_032185.1 | 2 | 623 | ||
| GenBank mRNA | AF045766.1 | - | 2 | 646 | ||
| BC064084.1 | - | 2 | 623 | 575313 | ||
| GQ981323.1 | - | 2 | 664 | |||
| GQ981327.1 | - | 2 | 726 | |||
More Information
Other Species Targeted
Set Membership: |
|

Gpr33
Genomic Map