siRNA Details
| Gene Symbol: Rhpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163465.1 | NP_001156937.1 | 7 | 774 | ||
| NM_008164.2 | NP_032190.2 | 7 | 774 | |||
| GenBank mRNA | AK143132.1 | - | 7 | 755 | ||
| AK220431.1 | - | 7 | 691 | |||
| BC052010.1 | - | 7 | 733 | 5717786 | ||
| U43194.1 | - | 7 | 668 | |||

Rhpn1
Genomic Map