siRNA Details
| Gene Symbol: Grin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008171.3 | NP_032197.3 | 4 | 842 | ||
| GenBank mRNA | AJ459261.1 | - | 4 | 843 | ||
| AJ459262.1 | - | 4 | 677 | |||
| AJ459263.1 | - | 3 | 604 | |||
| AK002963.1 | - | 4 | 843 | |||
| AK163195.1 | - | 572 | ||||
| D10651.2 | - | 1 | 79 | |||

Grin2b
Genomic Map