siRNA Details
| Gene Symbol: Grin2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008172.2 | NP_032198.2 | 9 | 2266 | ||
| GenBank mRNA | AB154244.1 | - | 9 | 2152 | ||
| AB154245.1 | - | 9 | 2152 | |||
| D12822.1 | - | 9 | 2152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grin2d| Gene Symbol: Grin2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008172.2 | NP_032198.2 | 9 | 2266 | ||
| GenBank mRNA | AB154244.1 | - | 9 | 2152 | ||
| AB154245.1 | - | 9 | 2152 | |||
| D12822.1 | - | 9 | 2152 | |||