siRNA Details
| Gene Symbol: H1f0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008197.3 | NP_032223.2 | 1 | 1861 | ||
| GenBank mRNA | AK051213.1 | - | 1 | 1624 | ||
| AK077473.1 | - | 1 | 1538 | |||
| AK090029.1 | - | 1 | 1629 | |||
| AK145675.1 | - | 1 | 1513 | |||
| AK146077.1 | - | 1 | 1624 | |||
| AK153990.1 | - | 1 | 1625 | |||
| AK155312.1 | - | 1 | 1556 | |||
| BC003830.1 | - | 1 | 1610 | 3600538 | ||
| BC011493.1 | - | 1 | 1608 | 4166167 | ||
| BC110361.1 | - | 1 | 1613 | 5343233 | ||

H1f0
Genomic Map