siRNA Details
| Gene Symbol: Mr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008209.4 | NP_032235.1 | 6 | 1216 | ||
| GenBank mRNA | AF010448.1 | - | 6 | 1127 | ||
| AF010449.1 | - | 6 | 942 | |||
| AF010450.1 | - | 6 | 942 | |||
| AF010451.1 | - | 3 | 1911 | |||
| AF010452.1 | - | 6 | 942 | |||
| AF010453.1 | - | 942 | ||||
| AK042119.1 | - | 6 | 1057 | |||
| AK137171.1 | - | 6 | 1057 | |||
| AK144040.1 | - | 6 | 1161 | |||
| AK162712.1 | - | 6 | 1173 | |||
| AK202684.1 | - | 1 | 39 | |||
| BC026137.1 | - | 6 | 1244 | 4017492 | ||

Mr1
Genomic Map