siRNA Details
| Gene Symbol: Has1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008215.2 | NP_032241.1 | 3 | 812 | ||
| GenBank mRNA | AK053726.1 | - | 2 | 2397 | ||
| AK138203.1 | - | 2 | 98 | |||
| BC125424.1 | - | 3 | 791 | 40129939 | ||
| BC125426.1 | - | 3 | 791 | 40129941 | ||
| D82964.1 | - | 3 | 818 | |||

Has1
Genomic Map