siRNA Details
| Gene Symbol: Has3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008217.4 | NP_032243.2 | 3 | 858 | ||
| GenBank mRNA | AK028582.1 | - | 3 | 802 | ||
| BC138152.1 | - | 3 | 754 | 8861171 | ||
| BC138161.1 | - | 3 | 753 | 8861180 | ||
| U86408.2 | - | 3 | 859 | |||

Has3
Genomic Map