siRNA Details
| Gene Symbol: Hells | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008234.3 | NP_032260.2 | 4 | 376 | ||
| GenBank mRNA | AF155210.1 | - | 4 | 376 | ||
| AK013266.1 | - | 4 | 415 | |||
| AK088901.1 | - | 2 | 1066 | |||
| AK147126.1 | - | 4 | 407 | |||
| AK166636.1 | - | 4 | 376 | |||
| BC090671.1 | - | 4 | 421 | |||
| BC100394.1 | - | 4 | 373 | 30480539 | ||
| U25691.2 | - | 4 | 409 | |||

Hells
Genomic Map