siRNA Details
| Gene Symbol: Hells | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008234.3 | NP_032260.2 | 14 | 1591 | ||
| GenBank mRNA | AF155210.1 | - | 14 | 1591 | ||
| AK013266.1 | - | 14 | 1630 | |||
| AK147126.1 | - | 14 | 1622 | |||
| AK166636.1 | - | 14 | 1592 | |||
| BC090671.1 | - | 14 | 1635 | |||
| U25691.2 | - | 14 | 1624 | |||

Hells
Genomic Map