siRNA Details
| Gene Symbol: Foxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277290.1 | NP_001264219.1 | 9 | 2201 | ||
| NM_008238.2 | NP_032264.1 | 9 | 2204 | |||
| GenBank mRNA | BC108980.1 | - | 8 | 2089 | 40047616 | |
| BC108981.1 | - | 8 | 2092 | 40047617 | ||
| X81593.1 | - | 9 | 2154 | |||

Foxn1
Genomic Map