siRNA Details
| Gene Symbol: Foxg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160112.1 | NP_001153584.1 | 1 | 3572 | ||
| NM_008241.2 | NP_032267.1 | 2 | 2534 | |||
| GenBank mRNA | AK132305.1 | - | 1 | 3560 | ||
| BC046958.1 | - | 1 | 2172 | 6314329 | ||
| BC055774.1 | - | 1 | 1866 | |||
| BC064449.1 | - | 1 | 2477 | |||
| BC079597.1 | - | 1 | 3238 | |||
| U36760.1 | - | 2 | 2533 | |||

Foxg1
Genomic Map