siRNA Details
| Gene Symbol: Hgs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159328.1 | NP_001152800.1 | 16 | 1516 | ||
| NM_008244.3 | NP_032270.3 | 16 | 1513 | |||
| GenBank mRNA | AK046299.1 | - | 16 | 1474 | ||
| AK145031.1 | - | 16 | 1436 | |||
| AK166441.1 | - | 16 | 1487 | |||
| BC003239.1 | - | 16 | 1457 | 3497966 | ||
| D50050.1 | - | 16 | 1463 | |||

Hgs
Genomic Map