siRNA Details
| Gene Symbol: Hmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008257.3 | NP_032283.3 | 1 | 113 | ||
| GenBank mRNA | BC139482.1 | - | 1 | 209 | 8862504 | |
| BC139483.1 | - | 1 | 209 | 8862505 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmx3| Gene Symbol: Hmx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008257.3 | NP_032283.3 | 1 | 113 | ||
| GenBank mRNA | BC139482.1 | - | 1 | 209 | 8862504 | |
| BC139483.1 | - | 1 | 209 | 8862505 | ||