siRNA Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008261.2 | NP_032287.2 | 10 | 2287 | ||
| GenBank mRNA | BC039220.1 | - | 10 | 2287 | 4238842 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnf4a| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008261.2 | NP_032287.2 | 10 | 2287 | ||
| GenBank mRNA | BC039220.1 | - | 10 | 2287 | 4238842 | |