siRNA Details
| Gene Symbol: Onecut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008262.3 | NP_032288.1 | 1 | 816 | ||
| GenBank mRNA | BC024053.1 | - | 1 | 816 | 5101697 | |
| U95945.1 | - | 1 | 609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Onecut1| Gene Symbol: Onecut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008262.3 | NP_032288.1 | 1 | 816 | ||
| GenBank mRNA | BC024053.1 | - | 1 | 816 | 5101697 | |
| U95945.1 | - | 1 | 609 | |||