siRNA Details
| Gene Symbol: Hoxb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008266.5 | NP_032292.3 | 1 | 394 | ||
| GenBank mRNA | AK017686.1 | - | 1 | 395 | ||
| BC103597.1 | - | 1 | 391 | 40046066 | ||
| BC103598.1 | - | 1 | 391 | 40046076 | ||
| BC103605.1 | - | 1 | 391 | 40046069 | ||
| BC103606.1 | - | 1 | 391 | 40046068 | ||

Hoxb1
Genomic Map