siRNA Details
| Gene Symbol: Hoxb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008268.2 | NP_032294.2 | 2 | 1777 | ||
| GenBank mRNA | AK133512.1 | - | 2 | 1693 | ||
| BC040755.1 | - | 3408 | ||||
| M26283.1 | - | 2 | 1777 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxb5| Gene Symbol: Hoxb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008268.2 | NP_032294.2 | 2 | 1777 | ||
| GenBank mRNA | AK133512.1 | - | 2 | 1693 | ||
| BC040755.1 | - | 3408 | ||||
| M26283.1 | - | 2 | 1777 | |||