siRNA Details
| Gene Symbol: Hoxd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008273.2 | NP_032299.1 | 2 | 919 | ||
| GenBank mRNA | AK054090.1 | - | 2 | 327 | ||
| BC138115.1 | - | 2 | 919 | 8861134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxd11| Gene Symbol: Hoxd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008273.2 | NP_032299.1 | 2 | 919 | ||
| GenBank mRNA | AK054090.1 | - | 2 | 327 | ||
| BC138115.1 | - | 2 | 919 | 8861134 | ||