siRNA Details
| Gene Symbol: Dnajb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008299.3 | NP_032325.2 | 1 | 659 | ||
| GenBank mRNA | AK005688.1 | - | 1 | 617 | ||
| AK005690.1 | - | 1 | 608 | |||
| BC048490.1 | - | 1 | 627 | 6775233 | ||
| U95607.1 | - | 1 | 661 | |||

Dnajb3
Genomic Map