siRNA Details
| Gene Symbol: Dnajb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008299.3 | NP_032325.2 | 1 | 853 | ||
| GenBank mRNA | AK005690.1 | - | 1 | 802 | ||
| U95607.1 | - | 1 | 855 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajb3| Gene Symbol: Dnajb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008299.3 | NP_032325.2 | 1 | 853 | ||
| GenBank mRNA | AK005690.1 | - | 1 | 802 | ||
| U95607.1 | - | 1 | 855 | |||