siRNA Details
| Gene Symbol: Ifi203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF022371.1 | - | 1175 | |||
| AK040214.1 | - | 7 | 1115 | |||
| AK137616.1 | - | 9 | 2500 | |||
| AK144663.1 | - | 8 | 1240 | |||
| AK161531.1 | - | 5 | 957 | |||
| AK164695.1 | - | 5 | 952 | |||
| AK169754.1 | - | 7 | 1109 | |||
| BB206743.2 | - | 2 | 324 | |||
| BC008167.1 | - | 1361 | 3257492 | |||
| L14559.1 | - | 4 | 751 | |||

Ifi203
Genomic Map