siRNA Details
| Gene Symbol: Ifit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008331.3 | NP_032357.2 | 2 | 753 | ||
| GenBank mRNA | AK089608.1 | - | 2 | 760 | ||
| AK131669.1 | - | 2 | 751 | |||
| AK144230.1 | - | 2 | 758 | |||
| AK150352.1 | - | 2 | 760 | |||
| AK150424.1 | - | 2 | 754 | |||
| AK150609.1 | - | 2 | 763 | |||
| AK151125.1 | - | 2 | 763 | |||
| AK151158.1 | - | 2 | 754 | |||
| AK151278.1 | - | 2 | 763 | |||
| AK151298.1 | - | 2 | 757 | |||
| AK151983.1 | - | 2 | 754 | |||
| AK152112.1 | - | 2 | 756 | |||
| AK152753.1 | - | 2 | 754 | |||
| AK152853.1 | - | 1 | 428 | |||
| AK153184.1 | - | 2 | 756 | |||
| AK153500.1 | - | 2 | 754 | |||
| AK153532.1 | - | 2 | 763 | |||
| AK157848.1 | - | 2 | 754 | |||
| BC003768.1 | - | 2 | 737 | 3496673 | ||
| CT010226.1 | - | 1 | 659 | |||
| U43084.1 | - | 2 | 728 | |||

Ifit1
Genomic Map