siRNA Details
| Gene Symbol: PWP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005049.2 | NP_005040.2 | 5 | 443 | ||
| XM_011529667.2 | XP_011527969.1 | 5 | 508 | |||
| GenBank mRNA | AK314184.1 | - | 5 | 447 | ||
| BC013309.2 | - | 5 | 353 | 3528790 | ||
| BC014988.1 | - | 5 | 353 | 4906611 | ||
| U53346.1 | - | 5 | 377 | |||
| U56085.1 | - | 5 | 388 | |||
| X95263.1 | - | 5 | 384 | |||
| Gene Symbol: LOC102724159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006723910.1 | XP_006723973.1 | 5 | 480 | ||

PWP2
Genomic Map