siRNA Details
| Gene Symbol: Il12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF128210.1 | - | 3 | 251 | ||
| AF128211.1 | - | 3 | 251 | |||
| AF128212.1 | - | 3 | 251 | |||
| AF128213.1 | - | 3 | 251 | |||
| AK153729.1 | - | 1 | 218 | |||
| BC116851.1 | - | 3 | 255 | |||
| BC116855.1 | - | 3 | 255 | |||
| M86672.1 | - | 3 | 353 | |||

Il12a
Genomic Map