siRNA Details
| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008353.2 | NP_032379.2 | 3 | 324 | ||
| GenBank mRNA | AK137577.1 | - | 3 | 325 | ||
| AK171937.1 | - | 3 | 301 | |||
| BC057212.1 | - | 3 | 307 | 5350447 | ||
| U23922.1 | - | 3 | 279 | |||

Il12rb1
Genomic Map