siRNA Details
| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC057212.1 | - | 5 | 562 | 5350447 | |
| U23922.1 | - | 5 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il12rb1| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC057212.1 | - | 5 | 562 | 5350447 | |
| U23922.1 | - | 5 | 534 | |||