siRNA Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008372.4 | NP_032398.3 | 6, 7 | 942 | ||
| GenBank mRNA | AF078906.1 | - | 6, 7 | 808 | ||
| AK040364.1 | - | 6, 7 | 925 | |||
| AK042264.1 | - | 6, 7 | 887 | |||
| AK154215.1 | - | 5, 6 | 756 | |||
| AK154516.1 | - | 6, 7 | 903 | |||
| BC089571.1 | - | 6, 7 | 894 | 6758487 | ||
| M29697.1 | - | 6, 7 | 855 | |||

Il7r
Genomic Map