siRNA Details
| Gene Symbol: Inhbe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008382.2 | NP_032408.2 | 1 | 408 | ||
| GenBank mRNA | BC010404.1 | - | 1 | 249 | 4160980 | |
| U96386.1 | - | 1 | 396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inhbe| Gene Symbol: Inhbe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008382.2 | NP_032408.2 | 1 | 408 | ||
| GenBank mRNA | BC010404.1 | - | 1 | 249 | 4160980 | |
| U96386.1 | - | 1 | 396 | |||