siRNA Details
Gene Symbol: Inpp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY007563.1 | - | 7 | 664 | ||
BC028864.1 | - | 7 | 650 | 4457437 | ||
CA315937.1 | - | 7 | 647 | |||
CK390048.1 | - | 3 | 288 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Inpp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY007563.1 | - | 7 | 664 | ||
BC028864.1 | - | 7 | 650 | 4457437 | ||
CA315937.1 | - | 7 | 647 | |||
CK390048.1 | - | 3 | 288 |