siRNA Details
| Gene Symbol: Edn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010104.3 | NP_034234.1 | 5 | 2141 | ||
| GenBank mRNA | AK040778.1 | - | 5 | 1800 | ||
| AK152647.1 | - | 5 | 1792 | |||
| AK153437.1 | - | 5 | 1797 | |||
| BC029547.1 | - | 5 | 1933 | 5059522 | ||
| U35233.1 | - | 5 | 1548 | |||

Edn1
Genomic Map