siRNA Details
| Gene Symbol: Eif4ebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010124.2 | NP_034254.1 | 3 | 1364 | ||
| GenBank mRNA | AK133912.1 | - | 3 | 1251 | ||
| AK147814.1 | - | 25 | ||||
| AK148374.1 | - | 1405 | ||||
| AK164011.1 | - | 299 | ||||
| BC015082.1 | - | 4 | 1345 | 4218820 | ||
| BC119539.2 | - | 4 | 1328 | 40056467 | ||
| U75530.1 | - | 3 | 1359 | |||

Eif4ebp2
Genomic Map