siRNA Details
| Gene Symbol: En2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010134.3 | NP_034264.1 | 2 | 1887 | ||
| GenBank mRNA | AK157766.1 | - | 2 | 1889 | ||
| L12705.1 | - | 2 | 1100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

En2| Gene Symbol: En2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010134.3 | NP_034264.1 | 2 | 1887 | ||
| GenBank mRNA | AK157766.1 | - | 2 | 1889 | ||
| L12705.1 | - | 2 | 1100 | |||