siRNA Details
| Gene Symbol: Ephb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159571.1 | NP_001153043.1 | 6 | 1483 | ||
| NM_010144.6 | NP_034274.4 | 6 | 1483 | |||
| GenBank mRNA | AK046884.1 | - | 6 | 1421 | ||
| AK049340.1 | - | 6 | 1484 | |||
| AK132392.1 | - | 6 | 1483 | |||
| AK143422.1 | - | 1453 | ||||
| AK199938.1 | - | 2 | 49 | |||
| BC013342.1 | - | 6 | 1143 | |||
| BC016505.1 | - | 6 | 1435 | 3709926 | ||
| BC090839.1 | - | 6 | 1260 | |||
| U06834.1 | - | 7 | 1096 | |||

Ephb4
Genomic Map